Level sensors
About this item
These level sensors provide dependable protection from overflow when filling barrels. The float positions itself securely at the opening by its own weight while the barrel fills up. Available in different materials, therefore suitable for all chemicals.
- Integrated ventilation for safe pressure equalisation
- Fill level control for containers up to 200 liters
Float for easy level control when filling/refuelling drums. Robust and chemically resistant for heavy-duty use. Made of dissipative HD-PE material and compatible for all barrels with thread G 3/4".
2 Options Available Below
Product Details & Documents
These level sensors provide dependable protection from overflow when filling barrels. The float positions itself securely at the opening by its own weight while the barrel fills up. Available in different materials, therefore suitable for all chemicals.
- Integrated ventilation for safe pressure equalisation
- Fill level control for containers up to 200 liters
Float for easy level control when filling/refuelling drums. Robust and chemically resistant for heavy-duty use. Made of dissipative HD-PE material and compatible for all barrels with thread G 3/4".
Documents
GHS Poster Important Lab and HPLC Safety Guidelines
Application Notes
Product Family Options
Product Information
- TypeLengthØAvailabilityPrice
- SCTV107883Level control for barrels with thread G3/4"
- SCTV107884Level control for barrels with thread G3/4"Level control for barrels with thread G3/4"Lance: PTFE, white; Floater: PFA, white; Display: PE, red150 mm18 mmLog in for availability1 * 1 ST699,00 €



