ContactUsLinkComponent
Bilirubin conjugate, ditaurate, disodium salt, Calbiochem®
Bilirubin conjugate, ditaurate, disodium salt, Calbiochem®
Catalog # 201102-100
CAS Number:  
Bilirubin conjugate, ditaurate, disodium salt, Calbiochem®
Catalog # 201102-100
CAS Number:  
Specifications
  • Pk:
    100 mg
  • Description:
    Bilirubin conjugate, ditaurate, disodium salt
Specifications
About this item
Bilirubin conjugate, ditaurate is suitable as a direct bilirubin standard. It behaves similarly to purified bilirubin glucuronides from human bile in diazo tests.
Note: Unstable in solution.