Orders LinkContactUsLinkComponent
Boot covers, Secutex® per, PP/PE
Boot covers, Secutex® per, PP/PE
Boot covers, Secutex® per, PP/PE
Certificates
Boot covers, Secutex® per, PP/PE
2 Options Available Below
Product Details & Documents
These boot covers provide protection against fine dusts and splashes of aggressive liquid chemicals. Partial resistance to splashing.
  • Smooth, lint-free and abrasion-resistant material (PP spunbond with PE laminate)
  • Permeable to air and water vapour
  • Impervious to particles, with partial resistance to splashing and also anti-static
  • Choice of boot covers either elasticated or with ties
Ideal for use in the remediation of contaminated sites, building reconstruction or in asbestos abatement as well as in spray work in agriculture or in industry and car paint shops.
*One size fits all.
Certifications: Cat. III, type 5 + 6
Specifications for Product Family
Specifications
  • Catalog No:
  • 113-0780
  • 113-0779
  • Garment application:
  • Chemical protection|Laboratory|Industrial
  • Chemical protection|Laboratory|Industrial
Specifications
Product Family Options
Product Information
  • Colour
    Shoe size
    Availability
    Price
  • 113-0780
    With ties
    With ties
    White
    U*
    Log in for availability
    Log in for availability
    1 * 150 PAIRE
    1 * 150 PAIRE - 381,00 €
    381,00 €
    1 * 150 PAIRE
       381,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-0780
    Garment application
    Chemical protection|Laboratory|Industrial
  • 113-0779
    Elasticated
    Elasticated
    White
    U*
    Log in for availability
    Log in for availability
    1 * 150 PAIRE
    1 * 150 PAIRE - 341,00 €
    341,00 €
    1 * 150 PAIRE
       341,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-0779
    Garment application
    Chemical protection|Laboratory|Industrial