Orders LinkContactUsLinkComponent
VWR® Forceps
VWR® Forceps
VWR® Forceps
Supplier:  VWR Collection
Certificates
VWR® Forceps
6 Options Available Below
Product Details & Documents
PTFE, white.
  • Very good chemical resistance, inert, non stick
  • Temperature resistant from –200 to +280 °C
  • Smooth surfaces prevent contamination and allow easy cleaning
Product Family Options
Product Information