Transfection reagents, DharmaFECT® siRNA
DHART-2001-03
:
- Pk:1,5 mL
- siRNA:Yes
- Description:DharmaFECT 1, 1.5 ml
A panel of four transfection reagents optimised for the efficient delivery of siRNA into mammalian cells.
- Specialised formulations for successful delivery of siRNA in a wide range of cell lines
- Effective delivery for target silencing at low siRNA concentrations
- Low cellular toxicity over a broad range of experimental conditions
- Free of microbial contaminants and ribonucleases
Each transfection reagent is provided as a liquid at a concentration of 1 mg/ml.