About this item
HyClone™ charcoal/dextran treated foetal bovine serum (FBS) is available for users requiring reduced levels of various hormones. High quality FBS is processed using a charcoal/dextran treatment that has been shown to reduce the levels of many hormones and growth factors.
- Sourced in the USA with complete traceability back to original source
- Low in antibodies and high in growth factors
- Produced using true pool technology for uniformity and consistency from bottle to bottle
- Filtered through three sequential 100nm (0.1 µm) pore size-rated filters
- Virus panel testing according to 9 CFR 113.53
HyClone charcoal/dextran treated Fetal Bovine Serum is available for users requiring reduced levels of various hormones. High-quality FBS is processed using a proprietary charcoal/dextran treatment that has been shown to reduce the levels of many hormones and growth factors. This serum is assayed for a variety of components before and after treatment to ensure the efficacy of the treatment. Charcoal/dextran treatment has been shown to be effective in reducing steroid levels for use of the serum in receptor studies or estrogen-related investigations. Other studies indicate that charcoal/dextran treatments may be used to minimize lot-to-lot serum variability.
Charcoal/Dextran Treated FBS is assayed for gamma globulin, alkaline phosphatase, lactate dehydrogenase, glutamic pyruvic transaminase, glutamic oxaloacetic transaminase, pH, total protein, albumin, blood urea nitrogen, creatinine, total bilirubin, sodium, potassium, calcium, chloride, inorganic phosphorus, osmolality, iron, total iron binding capacity (TIBC), percent iron saturation, glucose, and IgG. Included assays are subject to change without notice.
Additional treatments available including heat-inactivation and irradiation. Additional testing, including EMEA-conforming testing, is also available.