HyClone™ FetalClone™ I serum, U.S. origin
Supplier: HyClone products (Cytiva)
About this item
HyClone™ FetalClone® I is a foetal bovine serum alternative optimised for the growth of hybridomas with comparable IgG levels to those found in FBS.
- IgG levels comparable with those found in FBS
- Demonstrated cell growth performance equivalency to FBS
- High lot-to-lot consistency
- Low endotoxin and hemoglobin levels
Sterile filtered using three sequential 100 nm (0.1 µm) pore-size rated filters. Each lot undergoes extensive quality performance and analytical testing.
5 Options Available Below
Product Details & Documents
HyClone™ FetalClone® I is a foetal bovine serum alternative optimised for the growth of hybridomas with comparable IgG levels to those found in FBS.
- IgG levels comparable with those found in FBS
- Demonstrated cell growth performance equivalency to FBS
- High lot-to-lot consistency
- Low endotoxin and hemoglobin levels
Sterile filtered using three sequential 100 nm (0.1 µm) pore-size rated filters. Each lot undergoes extensive quality performance and analytical testing.
Documents
Certificates
Search by Lot NumberSpecifications for Product Family
Specifications
- Catalog No:
- SH30080.02
- HYCLSH30080.02HI
- HYCLSH30080.03
- HYCLSH30080.03HI
- SH30080.03IR25
- Storage temperature:
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
Specifications
Product Family Options
Product Information
- SterilitySerum formatAvailabilityPrice
- SH30080.02FetalClone® I, US originSpecifications:
More
Specifications
Cat. No.SH30080.02Storage temperatureSera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C. - HYCLSH30080.02HIFetalClone® I, US origin, heat inactivatedFetalClone® I, US origin, heat inactivatedSterile filteredFrozenLog in for availability1 * 100 mL109,00 €Specifications:
More
Specifications
Cat. No.HYCLSH30080.02HIStorage temperatureSera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C. - HYCLSH30080.03FetalClone® I, US originSpecifications:
More
Specifications
Cat. No.HYCLSH30080.03Storage temperatureSera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C. - HYCLSH30080.03HIFetalClone® I, US origin, heat inactivatedFetalClone® I, US origin, heat inactivatedSterile filteredFrozenLog in for availability1 * 500 mL370,00 €Specifications:
More
Specifications
Cat. No.HYCLSH30080.03HIStorage temperatureSera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C. - SH30080.03IR25FetalClone® I, US origin, irradiated 25 - 40 kGyFetalClone® I, US origin, irradiated 25 - 40 kGyGamma irradiatedFrozenLog in for availability1 * 500 mL370,00 €Specifications:
More
Specifications
Cat. No.SH30080.03IR25Storage temperatureSera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.