Order Entry
Belgium
ContactUsLinkComponent
HyClone Cell Boost™ Process Supplement Kit
HyClone Cell Boost™ Process Supplement Kit
Catalog # SH30890
CAS Number:  
undefined
HyClone Cell Boost™ Process Supplement Kit
Catalog # SH30890
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    HyClone Cell Boost™ process supplement kit
  • Storage temperature:
    2…8 °C
  • Pk Info:
    6×100 g
  • Form:
    Powder

Specifications

About this item

The HyClone Cell Boost™ process supplement kit contains 6×100 g samples of supplements designed to increase cell productivity in a variety of cell lines. Each supplement is designed to provide nutrients to fed-batch culture applications.

  • Supplements are chemically-defined and contain no animal components
  • Supplements have been tested in a variety of cell culture systems, including shaker flasks and bioreactors
  • Each supplement is also available in larger packaging configurations, please enquire for further information

Each Cell Boost formula is designed to provide a range of nutrients in novel combinations for multiple mammalian cell types, ideal for fed-batch applications or media customization. Each supplement has been tested in a variety of cell culture systems, including shaker flasks and bioreactors.