Order Entry
Belgium
ContactUsLinkComponent
VWR®, Taq DNA polymerase glycerol free
VWR®, Taq DNA polymerase glycerol free
Supplier:  VWR Chemicals
Certificates
undefined
VWR®, Taq DNA polymerase glycerol free
Supplier:  VWR Chemicals
Certificates

Specifications

  • Application:
    Freeze-drying
    Standard amplification
  • Format:
    Enzyme with/without buffer
  • High fidelity:
    No
  • Hot start:
    No
  • Storage conditions:
    – 20 °C (long term), + 4 °C (6 months)
  • Amplicon size:
    ≤ 5kb
  • Processivity:
    Standard
  • Exonuclease activity:
    5’→3’
  • Product overhang:
    3'A
  • Features:
    Glycerol free
  • Buffer composition:
    25 mM MgCl2
  • Concentration:
    5 U/µl
  • Description:
    Taq DNA Polymerase Glycerol Free 5U/µl 5.000 units
  • Size:
    10 x 0.1 ml
  • Units:
    5000

Specifications

Frequently Bought Together