ContactUsLinkComponent
Sequencing septa mats, Axygen®
Sequencing septa mats, Axygen®
N° de catalogue 731-4130
Fournisseur:  Corning
undefined
Sequencing septa mats, Axygen®
N° de catalogue 731-4130
Fournisseur:  Corning
Spécifications
  • Description:
    Sequencing septa mat
  • Pk:
    10
Spécifications
À propos de cet article
Silicone septa mat for capillary sequencing of 96-well PCR plates. Mat forms a tight seal on wells but is easily accessible through septa in each well. Ideal for work on ABI multi-capillary sequencing instruments.