Thioflavin T fluorescent dye, Calbiochem®, Sigma-Aldrich®
Catalog # 596200-500
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:318.86 g/mol
Storage Temperature:Ambient
CAS Number:2390-54-7
Formula:C₁₇H₁₉ClN₂S
MDL Number:MFCD00011944