About this item
The Proline Kryomats units are floor-standing, compact cooling thermostats and are fitted with the Command control head as standard. This features a back-lit, graphic LCD display with actual values in 0,1/0,01 or 0,001 °C resolution, data input via either cursor keys or numerical soft keys. The display provides clear text messages and graphical representation of sequential temperature curves and is menu-driven in four languages (German, English, French and Spanish). With large bath opening and volumes of up to 40 litres, this range is particularly suitable for internal applications.
- Temperature range from −90 to +200 °C
- Integrated opto-decoupled RS232/485 interface as standard
- Control head console can be detached and used as a remote control (up to 50 meters)
- Programmable control timer includes real-time clock with storage for up to 150 temperature/time segments, each with five programmes
- Adjustable recirculation nozzle inside the bath for optimised temperature homogeneity
All models can be supplied either in air or water-cooled versions with a powerful 4-stage Variopump. The electronically-controlled bath edge heating prevents moisture in the atmosphere from condensing and freezing at low temperatures. The sophisticated cooling units deliver extremely powerful cooling performance and are highly efficient even at low temperatures. With energy-saving digital cooling management function, the SmartCool system ensures that each mode of operation is run with the cooling performance it requires, rather than simply at maximum power. This saves up to 75 per cent of energy requirements by virtually ruling out the need to employ heating to balance out temperatures and by significantly reducing the heat lost to the surroundings.
Maximum pump capacities:
19 L/min, 0,5 bar
Related Information
TDS LAUDA Proline Kryomat RP 3090 C Cooling Thermostat L001654 TDS LAUDA Proline Kryomat RP 3090 CW Cooling Thermostat L001658 TDS LAUDA Proline Kryomat RP 4050 C Cooling Thermostat L001653 TDS LAUDA Proline Kryomat RP 4050 CW Cooling Thermostat L001657 TDS LAUDA Proline Kryomat RP 4090 C Cooling Thermostat L001655