À propos de cet article
PS, solid or with transparent base. Plates are available with different well shapes (round U-bottom, conical V-bottom, flat F-bottom or C-flat bottom with curved edges), in different colours, and with a choice of treated surfaces.
- All BRANDplates® are alphanumerically labelled, stackable and compatible with most standard microplate readers and washers
- Standard 96-well plates have coloured alphanumeric code identifying surface type (cell culture treated orange, immuno treated blue, non-treated grey)
- White plates provide maximum reflection and minimum crosstalk during luminescence measurements; black plates provide minimum back light scatter during fluorescence analyses
immunoGrade™: The surface of these plates is optimised for the immobilisation of IgG, showing a high binding capacity for molecules with hydrophilic and hydrophobic regions. Plates with an immunoGrade™ surface are the best choice for standard ELISA applications.
hydroGrade™: Strongly hydrophilic surface. Especially well suited for binding molecules with predominantly hydrophilic regions. Plates with a hydroGrade™ surface can be used in liquid phase assays with hydrophobic components.
lipoGrade™: Strongly hydrophobic (lipophilic) surface. Especially suitable for binding molecules with hydrophobic regions. Microplates with lipoGrade™ surfaces can be used in liquid phase assays with hydrophilic components.
Strongly hydrophilic surface. Especially well suited for binding molecules with predominantly hydrophilic regions. Plates with a hydroGrade™ surface can be used in liquid phase assays with hydrophobic components.