Orders LinkContactUsLinkComponent
Sterilisation Boxes for Pipettes
Sterilisation Boxes for Pipettes
Fournisseur:   BOCHEM
Sterilisation Boxes for Pipettes
Fournisseur:  BOCHEM
Certificates
Sterilisation Boxes for Pipettes
Sterilisation Boxes for Pipettes
5 Options Available Below
Product Details & Documents
Stainless steel, with square base, autoclavable.
  • Can be autoclaved at 121 °C and sterilised at 205 °C
Bottom and underside of the lid are coated in silicone to minimise the risk of damaging the pipettes during transport and sterilisation
Product Family Options
Product Information
  • W×D×H
    Disponibilité
    Prix
  • 612-1626
    Square base
    Square base
    70×70×202 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 134,00 €
    134,00 €
    1 * 1 ST
       134,00 €
  • 612-1627
    Square base
    Square base
    70×70×282 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 135,00 €
    135,00 €
    1 * 1 ST
       135,00 €
  • 612-1628
    Square base
    Square base
    70×70×332 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 163,00 €
    163,00 €
    1 * 1 ST
       163,00 €
  • 612-1629
    Square base
    Square base
    70×70×427 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 173,00 €
    173,00 €
    1 * 1 ST
       173,00 €
  • 612-1630
    Square base
    Square base
    70×70×482 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 206,00 €
    206,00 €
    1 * 1 ST
       206,00 €