Order Entry
Belgium
ContactUsLinkComponent
Poly-spoon, with hollow handle
Poly-spoon, with hollow handle
  HAMMHSN179-19
undefined
Poly-spoon, with hollow handle
  HAMMHSN179-19

 

  • Description:
    One spoon end, one spatula end, with hollow handle (Ø 8 mm)
  • Length:
    190 mm
  • Seq:
    09
  • Pk:
    1

 

 

Poly-spoon made of stainless-steel.

  • Double-ended: One spoon and one spatula end