Dissecting scissors, type La Grange
La Grange, double curved, saw edge, sharp/sharp, 115 mm
Catalog # HAMML-HSB047-11
Supplier: HAMMACHER KARL
Jump to:
Product Details & Documents
Dissecting scissors made of stainless steel.
Specifications
- Type:La Grange, double curved, saw edge, sharp/sharp
- Length:115 mm
- Seq:05