Orders LinkContactUsLinkComponent
Dissecting scissors, type Kelly
Dissecting scissors, type Kelly
Kelly, curved, saw edge, sharp/sharp, 160 mm
Catalog # HAMMHSB033-16
Supplier:   HAMMACHER KARL
Dissecting scissors, type Kelly
Catalog # HAMMHSB033-16
Supplier:  HAMMACHER KARL
Dissecting scissors, type Kelly
Dissecting scissors, type Kelly
Kelly, curved, saw edge, sharp/sharp, 160 mm
Price
107,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Dissecting scissors made of stainless steel.
Specifications
  • Type:
    Kelly, curved, saw edge, sharp/sharp
  • Length:
    160 mm
  • Seq:
    03