Order Entry
Belgium
ContactUsLinkComponent
 
Storage Boxes with Lid
  SEMA8202
 :  SEMADENI
Storage Boxes with Lid
  SEMA8202
 :  SEMADENI

 

  • Capacity:
    13 l
  • Colour:
    Transparent
  • Description:
    with blue lid
  • Material:
    PP
  • W×D×H:
    395×275×180 mm
  • Pk:
    1

 

 

Boxes made of PP.

  • Stackable and nestable
  • Lid can be used on both sides according to use

Robust and transparent storage boxes with convenient closing clips.