

Potassium hydroxide 29.5 - 30.5% in aqueous solution
Catalog # 310550.2500
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic container
- Pk:2,5 L
- Seq:5096
Specifications
About this item
:
:56.11 g/mol
UN:1814
ADR:8,II
:GHS07
:GHS05
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553