Orders LinkContactUsLinkComponent
Hardness tablets
Hardness tablets
Hardness tablets
Supplier:  Lovibond
Certificates
Hardness tablets
2 Options Available Below
Product Details & Documents
The speed test is based on reverse titration. After adding a reagent tablet to a calibrated test tube, the water sample is added slowly until the colour of the solution changes (e.g. from red to blue). The user can then obtain the result from the liquid level.
A Yes/No test tells the user whether a specific ingredient is present in the water and/or if its concentration is higher or lower than a defined level.
A Yes/No test tells the user whether a specific ingredient is present in the water and/or if its concentration is higher or lower than a defined level.
Documents
Product Family Options
Product Information
  • For unit
    Pk
    Availability
    Price
  • LOVI515590BT
    T-Hardness speed test tablets
    T-Hardness speed test tablets
    AF445 Minikit
    100
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 41,40 €
    41,40 €
    1 * 100 ST
       41,40 €
  • LOVI515361BT
    Hardness Yes/No tablets
    Hardness Yes/No tablets
    AF423 Minikit
    250 Tab.
    Log in for availability
    Log in for availability
    1 * 250 Tablet
    1 * 250 Tablet - 87,70 €
    87,70 €
    1 * 250 Tablet
       87,70 €