Order Entry
Belgium
ContactUsLinkComponent
Anti-ATF4 Rabbit Polyclonal Antibody
Anti-ATF4 Rabbit Polyclonal Antibody
Catalog # BSBTA00371
Supplier:  Boster Bio
undefined
Anti-ATF4 Rabbit Polyclonal Antibody
Catalog # BSBTA00371
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    ATF4
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Cyclic AMP-dependent transcription factor ATF-4(ATF4) detection. Tested with WB in Human.

Type: Primary
Antigen: ATF4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human