ContactUsLinkComponent
Inlet adaptor kit for dual column installation
Inlet adaptor kit for dual column installation
Catalog # REST27185
Supplier:  Restek
undefined
Inlet adaptor kit for dual column installation
Catalog # REST27185
Supplier:  Restek
Specifications
  • Description:
    Inlet adaptor kit for dual column installation
  • Pk:
    1
Specifications
About this item
Inlet adaptor kit for dual column installation ideal for Agilent capillary injectors, split/splitless fitting for capillary columns.
  • Split/splitless fitting that accepts standard, two-hole capillary ferrules
  • Easier to install capillary columns due to the nut protruding farther from the insulated injection port chamber
Delivery: Kit includes adaptor fitting, 1/16" capillary nut, gold-plated 1,2 mm ID dual Vespel® ring inlet seal, one 0,4 mm ID two-hole ferrule and one 0,5 mm ID two-hole ferrule.