Split/splitless injection port, EZ Twist Top®
Catalog # REST22177
Supplier: Restek
Specifications
- Description:EZ Twist Top® split/splitless injection port
- Pk:1
Specifications
About this item
This split/splitless injection port changes inlet liners faster and more easily.
- Gas lines are attached to shell weldment bottom to help eliminate broken gas lines
- Gas lines don’t interfere with routine maintenance
- Weldment stays secure in the tool for reattachment
Weldment removal tool allows for quick removal of the hot weldment without fingers ever touching hot surfaces.
Delivery: Kit includes weldment, shell weldment, 2 weldment O-rings, Siltek dual Vespel ring inlet seal, septum nut, reducing nut, stainless steel capillary nut for use with 1.6 mm ferrules and weldment removal tool.