ContactUsLinkComponent
Flexible inspection light
Flexible inspection light
Catalog # REST22627
Supplier:  Restek
undefined
Flexible inspection light
Catalog # REST22627
Supplier:  Restek
Specifications
  • Description:
    Flexible inspection light
  • Pk:
    1
Specifications
About this item
This flexible inspeciton light inspects inside surfaces of sample cylinders or other chambers.
  • Has a reach of 355 mm
  • LED life of 100000 hours