ContactUsLinkComponent
Make life easier (MLE) capillary tool kit for Thermo Scientific GCs
Make life easier (MLE) capillary tool kit for Thermo Scientific GCs
Catalog # REST22183
Supplier:  Restek
undefined
Make life easier (MLE) capillary tool kit for Thermo Scientific GCs
Catalog # REST22183
Supplier:  Restek
Specifications
  • Description:
    MLE capillary tool kit for Thermo Scientific GCs
  • Pk:
    1 KIT
Specifications
About this item
MLE tool kits conveniently provide the tools that make it easier to install and maintain capillary columns.
Delivery: Kit includes ¹/₈", ³/₁₆", ¹/₄" nylon brushes, ¹/₄", ³/₈", ³/₁₆" stainless steel wire tube brushes, stainless steel surface brush, 6 stainless steel jet reamers (0,25 to 0,65 mm OD), ¹/₄×⁵/₁₆" open end wrench, ³/₈×⁷/₁₆" open end wrench, rubber-tipped slide-lock tweezers, scoring wafers, inlet liner removal tool, septum puller, mini wool puller/inserter tool, 4-inch tapered needle file, swivel head flashlight, mini hand drill set, 15 cm compact steel ruler, pocket magnifier, high temperature string (1 m), pipe cleaner (12-inch), cotton tip swabs (pack of 25), capillary installation gauge, injector wrench, 6×7 mm open end wrench, 8×10 mm open end wrench and 16×17 mm open end wrench.