Orders LinkContactUsLinkComponent
Flow controllers, Veriflo
Flow controllers, Veriflo
Supplier:   Restek
Flow controllers, Veriflo
Supplier:  Restek
Certificates
Flow controllers, Veriflo
Flow controllers, Veriflo
25 Options Available Below
Product Details & Documents
Veriflo controllers can be used with passive air samplers in maintaining a constant flow rate.
Entire sample flow path is made of stainless steel or siltek® treated for maximum inertness.
Product Family Options
Product Information
  • Material
    Maximum Flow Rate
    Ø
    Availability
    Price
  • REST24239
    Flow controller, without orifice
    Flow controller, without orifice
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 350,00 €
    1 350,00 €
    1 * 1 ST
       1 350,00 €
  • REST24238
    Flow controller, without orifice
    Flow controller, without orifice
    Siltek® treated stainless steel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 490,00 €
    1 490,00 €
    1 * 1 ST
       1 490,00 €
  • REST24232
    Flow controller
    Flow controller
    Siltek® treated stainless steel
    0,5 – 2,0 ml/min
    0,02 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 620,00 €
    1 620,00 €
    1 * 1 ST
       1 620,00 €
  • REST24260
    Flow controller
    Flow controller
    Stainless steel
    2 – 4 ml/min
    0,03 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 280,00 €
    1 280,00 €
    1 * 1 ST
       1 280,00 €
  • REST24255
    Flow controller
    Flow controller
    Siltek® treated stainless steel
    2 – 4 ml/min
    0,03 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 650,00 €
    1 650,00 €
    1 * 1 ST
       1 650,00 €
  • REST24261
    Flow controller
    Flow controller
    Stainless steel
    4 – 8 ml/min
    0,041 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 400,00 €
    1 400,00 €
    1 * 1 ST
       1 400,00 €
  • REST24256
    Flow controller
    Flow controller
    Siltek® treated stainless steel
    4 – 8 ml/min
    0,041 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 650,00 €
    1 650,00 €
    1 * 1 ST
       1 650,00 €
  • REST24262
    Flow controller
    Flow controller
    Stainless steel
    8 – 15 ml/min
    0,051 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 370,00 €
    1 370,00 €
    1 * 1 ST
       1 370,00 €
  • REST24257
    Flow controller
    Flow controller
    Siltek® treated stainless steel
    8 – 15 ml/min
    0,051 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 650,00 €
    1 650,00 €
    1 * 1 ST
       1 650,00 €
  • REST24263
    Flow controller
    Flow controller
    Stainless steel
    15 – 30 ml/min
    0,076 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 370,00 €
    1 370,00 €
    1 * 1 ST
       1 370,00 €
  • REST24258
    Flow controller
    Flow controller
    Siltek® treated stainless steel
    15 – 30 ml/min
    0,076 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 650,00 €
    1 650,00 €
    1 * 1 ST
       1 650,00 €
  • REST24264
    Flow controller
    Flow controller
    Stainless steel
    30 – 80 ml/min
    0,152 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 370,00 €
    1 370,00 €
    1 * 1 ST
       1 370,00 €
  • REST24259
    Flow controller
    Flow controller
    Siltek® treated stainless steel
    30 – 80 ml/min
    0,152 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 650,00 €
    1 650,00 €
    1 * 1 ST
       1 650,00 €
Replacement orifices
Product Information
  • Material
    Maximum Flow Rate
    Ø
    Availability
    Price
  • REST24218
    Replacement orifice
    Replacement orifice
    Stainless steel
    0,5 – 2 ml/min
    0,02 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 220,00 €
    220,00 €
    1 * 1 ST
       220,00 €
  • REST24219
    Replacement orifice
    Replacement orifice
    Siltek® treated stainless steel
    0,5 – 2 ml/min
    0,02 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 340,00 €
    340,00 €
    1 * 1 ST
       340,00 €
  • REST24245
    Replacement orifice
    Replacement orifice
    Stainless steel
    2 – 4 ml/min
    0,03 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 220,00 €
    220,00 €
    1 * 1 ST
       220,00 €
  • REST24233
    Replacement orifice
    Replacement orifice
    Siltek® treated stainless steel
    2 – 4 ml/min
    0,03 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 340,00 €
    340,00 €
    1 * 1 ST
       340,00 €
  • REST24246
    Replacement orifice
    Replacement orifice
    Stainless steel
    4 – 8 ml/min
    0,041 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 220,00 €
    220,00 €
    1 * 1 ST
       220,00 €
  • REST24234
    Replacement orifice
    Replacement orifice
    Siltek® treated stainless steel
    4 – 8 ml/min
    0,041 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 340,00 €
    340,00 €
    1 * 1 ST
       340,00 €
  • REST24235
    Replacement orifice
    Replacement orifice
    Siltek® treated stainless steel
    8 – 15 ml/min
    0,051 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 340,00 €
    340,00 €
    1 * 1 ST
       340,00 €
  • REST24247
    Replacement orifice
    Replacement orifice
    Stainless steel
    8 – 15 ml/min
    0,051 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 220,00 €
    220,00 €
    1 * 1 ST
       220,00 €
  • REST24236
    Replacement orifice
    Replacement orifice
    Siltek® treated stainless steel
    15 – 30 ml/min
    0,076 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 340,00 €
    340,00 €
    1 * 1 ST
       340,00 €
  • REST24248
    Replacement orifice
    Replacement orifice
    Stainless steel
    15 – 30 ml/min
    0,076 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 220,00 €
    220,00 €
    1 * 1 ST
       220,00 €
  • REST24237
    Replacement orifice
    Replacement orifice
    Siltek® treated stainless steel
    30 – 80 ml/min
    0,152 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 340,00 €
    340,00 €
    1 * 1 ST
       340,00 €
  • REST24249
    Replacement orifice
    Replacement orifice
    Stainless steel
    30 – 80 ml/min
    0,152 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 220,00 €
    220,00 €
    1 * 1 ST
       220,00 €