ContactUsLinkComponent
Micropacked inlet conversion kits
Micropacked inlet conversion kits
Catalog # REST22426
Supplier:  Restek
undefined
Micropacked inlet conversion kits
Catalog # REST22426
Supplier:  Restek
Specifications
  • Description:
    Injection port adapter kit
  • Pk:
    1
Specifications
About this item
Micropacked inlet conversion kits are used to convert a capillary GC split/splitless inlet for use with ¹/₁₆" Ext.Ø micropacked columns.
  • For use with Agilent 5890 6890 and 7890 GCs
  • Sample pathways deactivated for ultimate inertness
Delivery: Kit includes dual large bore Vespel ring inlet seal, large bore reducing nut, large bore FID adaptor, ¹/₁₆" Vespel/graphite ferrule, ¹/₁₆" stainless steel nut, 4 mm splitless liner (intermediate polarity deactivated).