ContactUsLinkComponent
Injector wrench for Shimadzu
Injector wrench for Shimadzu
Catalog # REST21159
Supplier:  Restek
undefined
Injector wrench for Shimadzu
Catalog # REST21159
Supplier:  Restek
Specifications
  • Description:
    Injector wrench
  • Material:
    Stainless steel
  • Pk:
    1
Specifications
About this item
This stainless steel inlet wrench is ideal for Shimadzu 17A, 2010, and 2014 GCs.
  • Designed specifically for removing Shimadzu injection ports