Digital Solutions
Applications
Support
About
Belgium
ContactUsLinkComponent
Seals
Sealing Rings
O-rings for PerkinElmer PSS injector, Viton®
O-rings for PerkinElmer PSS injector, Viton®
REST20366
:
Restek
O-rings for PerkinElmer PSS injector, Viton®
REST20366
:
Restek
Description:
O-rings for liners with OD 4 mm
Material:
Viton®
Pk:
10