Orders LinkContactUsLinkComponent
Mobile Boxes for Hazardous Materials
Supplier:   CEMO
Mobile Boxes for Hazardous Materials
Supplier:  CEMO
Certificates
Mobile Boxes for Hazardous Materials
Mobile Boxes for Hazardous Materials
5 Options Available Below
Product Details & Documents
These mobile boxes with its stable design enables clean and safe storage of hazardous solid goods belonging to packing groups II or III and is also approved for transport according to ADR, RID and IMDG codes.
  • Easy to handle, with coloured lids
  • Specially shaped lid enables storage in two-layers
  • Up to eight boxes can be safely stacked on a Euro pallet
  • Beading in the container wall enables emptying with a rotary forklift
Product Family Options
Product Information
  • Capacity
    W×D×H
    Availability
    Price
  • CEMG7719
    Mobile box blue, with green lid
    Mobile box blue, with green lid
    170 l
    600×400×880 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 220,00 €
    220,00 €
    1 * 1 ST
       220,00 €
  • CEMG7720
    Mobile box blue, with blue lid
    Mobile box blue, with blue lid
    170 l
    600×400×880 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 298,00 €
    298,00 €
    1 * 1 ST
       298,00 €
  • CEMG7805
    Mobile box blue, with red lid
    Mobile box blue, with red lid
    250 l
    600×600×890 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 362,00 €
    362,00 €
    1 * 1 ST
       362,00 €
  • CEMG7806
    Mobile box blue, with blue lid
    Mobile box blue, with blue lid
    250 l
    600×600×890 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 362,00 €
    362,00 €
    1 * 1 ST
       362,00 €
  • CEMG8318
    Mobile box blue, with green lid
    Mobile box blue, with green lid
    250 l
    600×600×890 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 362,00 €
    362,00 €
    1 * 1 ST
       362,00 €