Orders LinkContactUsLinkComponent
Injector adapters
Injector adapters
Supplier:   Restek
Injector adapters
Supplier:  Restek
Certificates
Injector adapters
Injector adapters
2 Options Available Below
Product Details & Documents
These injector adapters made of stainless steel are ideal for PerkinElmer CAP injector.
Siltek®-treated version available for increased inertness.
Product Family Options
Product Information
  • For
    Availability
    Price
  • REST22318
    Injector adapter for PerkinElmer CAP injector, stainless steel, for PE-style capillary nuts
    Injector adapter for PerkinElmer CAP injector, stainless steel, for PE-style capillary nuts
    PE-style capillary nuts
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 152,00 €
    152,00 €
    1 * 1 ST
       152,00 €
  • REST22319
    Injector adapter for PerkinElmer CAP injector, Siltek®-treated, for 1/16" compression-style nuts
    Injector adapter for PerkinElmer CAP injector, Siltek®-treated, for 1/16" compression-style nuts
    1/16" compression-style nuts
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST