Order Entry
Belgium
ContactUsLinkComponent
Ferrules for Shimadzu 17A GCs
Ferrules for Shimadzu 17A GCs
Catalog # REST21121
Supplier:  Restek
undefined
Ferrules for Shimadzu 17A GCs
Catalog # REST21121
Supplier:  Restek

Specifications

  • Description:
    Graphite ferrules, 5 mm
  • Pk:
    10

Specifications

About this item

Ferrules with graphite construction for use with packed columns.