Orders LinkContactUsLinkComponent
Eudiometer tubes
Eudiometer tubes
Eudiometer tube, complete, includes with reservoir, leveling plate and PVC connecting hose, 1000 ml, 45
Catalog # NEUB2591-10-509
Eudiometer tubes
Catalog # NEUB2591-10-509
Eudiometer tubes
Eudiometer tube, complete, includes with reservoir, leveling plate and PVC connecting hose, 1000 ml, 45
Price
1 770,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Eudiometer tubes with spindle tap are suitable for determining the digested behaviour of sludge and biodegradability under anaerobic conditions.
  • Made of borosilicate glass 3.3
  • With reservoir and leveling plate
  • With PVC connecting hose
Certifications: According to DIN 38414 part 8.
Specifications
  • NS:
    45
  • Capacity:
    1000 ml
  • Description:
    Eudiometer tube, complete, includes with reservoir, leveling plate and PVC connecting hose
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.