Order Entry
Belgium
ContactUsLinkComponent
Human recombinant Stem Cell Factor (from E. coli), MP Biomedicals
Human recombinant Stem Cell Factor (from E. coli), MP Biomedicals
Catalog # ICNA0215869802
Supplier:  MP Biomedicals
CAS Number:  
Human recombinant Stem Cell Factor (from E. coli), MP Biomedicals
Catalog # ICNA0215869802
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    2 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    01
  • Storage conditions:
    –20 °C
  • Form:
    Solid
  • Protein/peptide name:
    Stem Cell Factor
  • Purity:
    >98%
  • Molecular weight:
    18.4 kD
  • Colour:
    White

Specifications

About this item

Stem Cell Factor (SCF) is a hematopoietic growth factor that exerts its activity at the early stages of hematopoiesis. It stimulates the proliferation of myeloid, erythroid, and lymphoid progenitors in bone marrow cultures and has been shown to act synergistically with colony stimulating factors.

  • Soluble in water and most aqueous buffers
  • Specific activity: ≤20 ng/ml