Orders LinkContactUsLinkComponent
Sump Pallets
Supplier:   CEMO
Sump Pallets
Supplier:  CEMO
Certificates
Sump Pallets
10 Options Available Below
Product Details & Documents
These small container/laboratory trays can be combined to cover a complete storage area and also can be used as storage/filling station. Trays can be used with or without perforated plate.
  • Made of polyethylene
  • With general construction inspection approval (application for admission)
  • Stackable and provides space-saving storage/transport
  • Fully recyclable and maintenance friendly
  • Rugged, removable PE grating
  • Can be connected to form a surface protection system
Stable PE access ramp with an integrated fixing bolt for connection to the PE grating, slight inclination for easy loading of the floor elements.
Product Family Options
Product Information
  • W×D×H
    Availability
    Price
  • CEMG10704
    Sump pallet, PE, 25 l, weight 1,8 kg
    Sump pallet, PE, 25 l, weight 1,8 kg
    600×400×165 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 71,20 €
    71,20 €
    1 * 1 ST
       71,20 €
  • CEMG10705
    Sump pallet, PE, 25 l with PE grating, load capacity 125 kg, weight 3,7 kg
    Sump pallet, PE, 25 l with PE grating, load capacity 125 kg, weight 3,7 kg
    600×400×180 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 196,00 €
    196,00 €
    1 * 1 ST
       196,00 €
  • CEMG10706
    Sump pallet, PE, 35 l, weight 2,2 kg
    Sump pallet, PE, 35 l, weight 2,2 kg
    800×400×165 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 133,00 €
    133,00 €
    1 * 1 ST
       133,00 €
  • CEMG10707
    Sump pallet, PE, 35 l with PE grating, load capacity 160 kg, weight 4,8 kg
    Sump pallet, PE, 35 l with PE grating, load capacity 160 kg, weight 4,8 kg
    800×400×180 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 218,00 €
    218,00 €
    1 * 1 ST
       218,00 €
  • CEMG10708
    Sump pallet, PE, 60 l, weight 3,0 kg
    Sump pallet, PE, 60 l, weight 3,0 kg
    800×600×165 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 122,00 €
    122,00 €
    1 * 1 ST
       122,00 €
  • CEMG10709
    Sump pallet, PE, 60 l with PE grating, load capacity 250 kg, weight 6,8 kg
    Sump pallet, PE, 60 l with PE grating, load capacity 250 kg, weight 6,8 kg
    800×600×180 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 243,00 €
    243,00 €
    1 * 1 ST
       243,00 €
  • CEMG10710
    Sump pallet, PE, 120 l, weight 5,2 kg
    Sump pallet, PE, 120 l, weight 5,2 kg
    1200×800×165 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 169,00 €
    169,00 €
    1 * 1 ST
       169,00 €
  • CEMG10711
    Sump pallet, PE,120 l with PE grating, load capacity 500 kg, weight 12,8 kg
    Sump pallet, PE,120 l with PE grating, load capacity 500 kg, weight 12,8 kg
    1200×800×180 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 322,00 €
    322,00 €
    1 * 1 ST
       322,00 €
  • CEMG10712
    Sump pallet, PE, 250 l, weight 10,5 kg
    Sump pallet, PE, 250 l, weight 10,5 kg
    1600×1200×165 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 263,00 €
    263,00 €
    1 * 1 ST
       263,00 €
  • CEMG10713
    Sump pallet, PE, 250 l with PE grating, load capacity 1000 kg, weight 25,7 kg
    Sump pallet, PE, 250 l with PE grating, load capacity 1000 kg, weight 25,7 kg
    1600×1200×180 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 684,00 €
    684,00 €
    1 * 1 ST
       684,00 €
Accessories
Product Information
  • Catalog #
    W×D×H
    Availability
    Price
  • Cross connector, short, for ground protection system
    Cross connector, short, for ground protection system
    CEMG10723
    115×115×35 mm
    Log in for availability
    1 * 1 ST - 18,10 €
Product Information
  • Catalog #
    W×D×H
    Availability
    Price
  • Connecting rail, short, for ground protection system
    Connecting rail, short, for ground protection system
    CEMG10722
    1540×15×15 mm
    Log in for availability
    1 * 1 ST - 29,90 €
Product Information
  • Catalog #
    W×D×H
    Availability
    Price
  • Access ramp, for ground protection system, load capacity 300 kg, weight 13 kg
    Access ramp, for ground protection system, load capacity 300 kg, weight 13 kg
    CEMG10714
    1260×800×200 mm
    Log in for availability
    1 * 1 ST - 308,00 €
Product Information
  • Catalog #
    W×D×H
    Availability
    Price
  • Connecting rail, long, for ground protection system
    Connecting rail, long, for ground protection system
    CEMG10721
    1140×15×15 mm
    Log in for availability
    1 * 1 ST - 23,80 €