Orders LinkContactUsLinkComponent
Sterilisation Boxes for Pipettes
Sterilisation Boxes for Pipettes
Supplier:   Brand
Sterilisation Boxes for Pipettes
Supplier:  Brand
Certificates
Sterilisation Boxes for Pipettes
Sterilisation Boxes for Pipettes
4 Options Available Below
Product Details & Documents
These pipette boxes made of aluminum are ideal for the storage and sterilisation of pipettes with different lengths.
  • With inner silicone coating
  • Very good temperature resistance
  • Very good chemical resistance
Product Family Options
Product Information
  • Availability
    Price
  • BRND28920
    Pipette box for the storage of pipettes (150 - 200 mm)
    Pipette box for the storage of pipettes (150 - 200 mm)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 64,40 €
    64,40 €
    1 * 1 ST
       64,40 €
  • BRND28925
    Pipette box for the storage of pipettes (235 - 305 mm)
    Pipette box for the storage of pipettes (235 - 305 mm)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 70,50 €
    70,50 €
    1 * 1 ST
       70,50 €
  • BRND28930
    Pipette box for the storage of pipettes (315 - 385 mm)
    Pipette box for the storage of pipettes (315 - 385 mm)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 76,80 €
    76,80 €
    1 * 1 ST
       76,80 €
  • BRND28935
    Pipette box for the storage of pipettes (315 - 485 mm)
    Pipette box for the storage of pipettes (315 - 485 mm)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 81,50 €
    81,50 €
    1 * 1 ST
       81,50 €