Orders LinkContactUsLinkComponent
Particle traps for use with PLOT columns
Particle traps for use with PLOT columns
Supplier:   Agilent
Particle traps for use with PLOT columns
Supplier:  Agilent
Certificates
Particle traps for use with PLOT columns
8 Options Available Below
Product Details & Documents
Though highly stabilised, it is impossible to guarantee that no particles will dislodge from the column wall. When used in valve-switching applications, the use of a particle trap can prevent scarring of the rotors.
Product Family Options
Particle traps for use with PLOT columns
Product Information
  • Availability
    Price
  • HEWL5181-3351
    Particle trap for use with PLOT columns, Int.Ø×L: 0,32 mm × 2,5 m
    Particle trap for use with PLOT columns, Int.Ø×L: 0,32 mm × 2,5 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 281,00 €
    281,00 €
    1 * 1 ST
       281,00 €
  • HEWL5181-3352
    Particle trap for use with PLOT columns, Int.Ø×L: 0,53 mm × 2,5 m
    Particle trap for use with PLOT columns, Int.Ø×L: 0,53 mm × 2,5 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 353,00 €
    353,00 €
    1 * 1 ST
       353,00 €
Particle traps for PoraPLOT columns
Product Information
  • Availability
    Price
  • VARICP4016
    Particle trap for PoraPLOT columns, fused silica, Int.Ø×L: 0,32 mm × 2,5 m
    Particle trap for PoraPLOT columns, fused silica, Int.Ø×L: 0,32 mm × 2,5 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 153,00 €
    153,00 €
    1 * 1 ST
       153,00 €
  • VARICP4017
    Particle trap for PoraPLOT columns, fused silica, Int.Ø×L: 0,53 mm × 2,5 m
    Particle trap for PoraPLOT columns, fused silica, Int.Ø×L: 0,53 mm × 2,5 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 193,00 €
    193,00 €
    1 * 1 ST
       193,00 €
  • VARICP4018
    Particle trap for PoraPLOT columns, UltiMetal, includes CP-UltiMetal connector, Int.Ø×L: 0,53 mm × 2,5 m
    Particle trap for PoraPLOT columns, UltiMetal, includes CP-UltiMetal connector, Int.Ø×L: 0,53 mm × 2,5 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 249,00 €
    249,00 €
    1 * 1 ST
       249,00 €
Particle trap connectors for PoraPLOT columns
Product Information
  • Availability
    Price
  • VARICP4788
    Particle trap connectors for PoraPLOT columns, fused silica, 0,25/0,32 mm ID
    Particle trap connectors for PoraPLOT columns, fused silica, 0,25/0,32 mm ID
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 227,00 €
    227,00 €
    1 * 10 ST
       227,00 €
  • VARICP4789
    Particle trap connectors for PoraPLOT columns, fused silica, 0,53 mm ID
    Particle trap connectors for PoraPLOT columns, fused silica, 0,53 mm ID
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 227,00 €
    227,00 €
    1 * 10 ST
       227,00 €
  • VARICP4795
    Particle trap connectors for PoraPLOT columns, UltiMetal, 0,25 mm ID
    Particle trap connectors for PoraPLOT columns, UltiMetal, 0,25 mm ID
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST