Catalase (from Aspergillus niger)
Catalog # 219261-100
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pk:100 KU
- CAS No.:9001-05-2
- Source:Aspergillus niger
- Storage conditions:−20 °C
- Form:Lyophilized powder
- Enzyme name:Catalase
Specifications
About this item
Catalase, fungal suspension from from Aspergillus niger is long-acting, extremely stable form of catalase that is active over a wide pH range (pH 1 to 12). Composed of four protein subunits, each containing a heme group bound to its active site.
Catalase is used for the removal of peroxides, the generation of oxygen and, in coupled systems, for the determination of metabolites e.g. uric acid. A very stable preparation particularly suitable for immobilisation.