Peristaltic pump heads, rotarus®
Supplier: HIRSCHMANN
About this item
5 Options Available Below
Product Details & Documents
Documents
Hirschmann brochure insert sheets rotarus® -pump series- tubes Hirschmann brochure rotarus® -pump series-
Product Literature
Hirschmann data sheets rotarus® peristaltic pump head MKF12-8 Hirschmann data sheets rotarus® peristaltic pump head MKF4-8 Hirschmann data sheets rotarus® peristaltic pump head MKF8-4 Hirschmann data sheets rotarus® peristaltic pump head PK10-16 Hirschmann data sheets rotarus® peristaltic pump head PK10-24
Datasheet
Specifications for Product Family
Specifications
- Catalog No:
- 224-0134
- 224-0135
- 224-0121
- 224-0122
- 224-0123
- Model:
- PK 10-16
- PK 10-24
- MKF 4-8
- MKF 8-4
- MKF 12-8
- Min. flow rate:
- 0,08 ml/min
- 0,66 ml/min
- 0,0009 ml/min
- 0,014 ml/min
- 0,0001 ml/min
- No. of rollers:
- 3
- 3
- 8
- 4
- 8
Specifications
Product Family Options
Product Information
- Maximum Flow RateAvailabilityPrice
- 224-0134Pump head, rotarus® PK 10-16, 1 channel, for tubes with inner diameter 1,6 - 11,1 mmPump head, rotarus® PK 10-16, 1 channel, for tubes with inner diameter 1,6 - 11,1 mm5,400 ml/minLog in for availability1 * 1 ST1 390,00 €Specifications:
More
Specifications
Cat. No.224-0134ModelPK 10-16Min. flow rate0,08 ml/minNo. of rollers3 - 224-0135Pump head, rotarus® PK 10-24, 1 channel, for tubes with inner diameter 4,8 - 9,5 mmPump head, rotarus® PK 10-24, 1 channel, for tubes with inner diameter 4,8 - 9,5 mm5,200 ml/minLog in for availability1 * 1 ST1 390,00 €Specifications:
More
Specifications
Cat. No.224-0135ModelPK 10-24Min. flow rate0,66 ml/minNo. of rollers3 - 224-0121Pump head, rotarus® MKF 4-8, 4 channels, for stopper tubesPump head, rotarus® MKF 4-8, 4 channels, for stopper tubes82,1 ml/minLog in for availability1 * 1 ST1 800,00 €Specifications:
More
Specifications
Cat. No.224-0121ModelMKF 4-8Min. flow rate0,0009 ml/minNo. of rollers8 - 224-0122Pump head, rotarus® MKF 8-4, 8 channels, for stopper tubesPump head, rotarus® MKF 8-4, 8 channels, for stopper tubes228 ml/minLog in for availability1 * 1 ST2 830,00 €Specifications:
More
Specifications
Cat. No.224-0122ModelMKF 8-4Min. flow rate0,014 ml/minNo. of rollers4 - 224-0123Pump head, rotarus® MKF 12-8, 12 channels, for stopper tubesPump head, rotarus® MKF 12-8, 12 channels, for stopper tubes27,2 ml/minLog in for availability1 * 1 ST4 370,00 €Specifications:
More
Specifications
Cat. No.224-0123ModelMKF 12-8Min. flow rate0,0001 ml/minNo. of rollers8