Gas-tight syringes, PTFE tip (SGE)
Fournisseur: Restek
Certificats
À propos de cet article
These gas-tight syringes with interchangeable barrels and plungers and tips extend performance and increase cost effectiveness.
- Suitable for gases and liquids
- High accuracy of dispensed volumes
Informations de commande: Needles have to be ordered separately for syringes without needles.