Micro-volume syringes, positive displacement, (SGE)
Fournisseur: Restek
Certificats
À propos de cet article
These micro-volume syringes have samples contained in the needle with full plunger displacement, ensuring no dead volume.
- High precision and accuracy down to 0,1 µl
- Seal between plunger and needle can be tightened for high-pressure injection
* 70 mm needle length, 0,47 mm OD, on-column syringe.
Informations de commande: Replaceable needle/plunger kits and barrels are available on request.
Replacement plunger-in-needle kits
Informations sur le produit
Frequently Bought Together