ContactUsLinkComponent
Thermal transfer printer labels, type B-351, Defender
Thermal transfer printer labels, type B-351, Defender
Catalog # BRDY020822
Supplier:  Brady
undefined
Thermal transfer printer labels, type B-351, Defender
Catalog # BRDY020822
Supplier:  Brady
Specifications
  • Type:
    B-351
  • Colour:
    White
  • Description:
    Printer labels
  • Dimensions:
    50×25 mm
  • Material:
    Vinyl
  • Model:
    THT-17-7351-3
  • Packed:
    3000 labels/roll
  • Pk:
    1 Roll
  • Adhesive:
    Permanent acrylic
Specifications
About this item
Printable, tamper-evident vinyl labels for thermal transfer printers.
  • Fractures easily
  • Prevents one-piece removal
Labels are designed to fracture easily in order to show signs of product tampering and to prevent one-piece label removal. Use caution when removing from liner as material is fragile.