Orders LinkContactUsLinkComponent
Capillary column caps
Supplier:   Restek
Capillary column caps
Supplier:  Restek
Certificates
Capillary column caps
Capillary column caps
2 Options Available Below
Product Details & Documents
These non-reusable column caps increase column lifetime, prevent moisture and air from entering the column during storage.
  • Attach to the column in seconds to form an airtight seal
  • Available in two styles, glass or silicone material
  • Glass caps are colour coded for identifying detector and injector ends
Product Family Options
Product Information
  • Availability
    Price
  • REST21044
    Capillary column caps, glass
    Capillary column caps, glass
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 142,00 €
    142,00 €
    1 * 20 ST
       142,00 €
  • REST22858
    Capillary column caps, silicone
    Capillary column caps, silicone
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 44,30 €
    44,30 €
    1 * 10 ST
       44,30 €