Orders LinkContactUsLinkComponent
Group lockout hasps, labelled
Group lockout hasps, labelled
Supplier:   Brady
Group lockout hasps, labelled
Supplier:  Brady
Certificates
Group lockout hasps, labelled
Group lockout hasps, labelled
12 Options Available Below
Product Details & Documents
These labelled group lockout hasps are constructed from tough 5052 anodised aluminium alloy.
  • 6 padlocks can be applied
  • Include tamperproof locking tabs
  • Write-on label surface accept pencils, pen or marker
  • Pen and pencil markings can be easily erased, allowing repeated use
  • Lock hole diameter: 8 mm
Delivery information: The assorted pack contains one each of the following colours: red, silver, blue, green and gold.
Product Family Options
Product Information
  • Colour
    W×D×H
    Availability
    Price
  • Blue
    76×5×180 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 125,00 €
    125,00 €
    1 * 5 ST
       125,00 €
  • Gold
    76×5×180 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 125,00 €
    125,00 €
    1 * 5 ST
       125,00 €
  • Green
    76×5×180 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 125,00 €
    125,00 €
    1 * 5 ST
       125,00 €
  • Red
    76×5×180 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 125,00 €
    125,00 €
    1 * 5 ST
       125,00 €
  • Silver
    76×5×180 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 125,00 €
    125,00 €
    1 * 5 ST
       125,00 €
  • Assorted colours
    76×5×180 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 125,00 €
    125,00 €
    1 * 5 ST
       125,00 €
  • Blue
    76×5×180 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 125,00 €
    125,00 €
    1 * 5 ST
       125,00 €
  • Gold
    76×5×180 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 125,00 €
    125,00 €
    1 * 5 ST
       125,00 €
  • Green
    76×5×180 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 125,00 €
    125,00 €
    1 * 5 ST
       125,00 €
  • Red
    76×5×180 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 125,00 €
    125,00 €
    1 * 5 ST
       125,00 €
  • Silver
    76×5×180 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 125,00 €
    125,00 €
    1 * 5 ST
       125,00 €
  • Assorted colours
    76×5×180 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 125,00 €
    125,00 €
    1 * 5 ST
       125,00 €