ContactUsLinkComponent
Acetate plate seal, MP Biomedicals
Acetate plate seal, MP Biomedicals
Catalog # ICNA097640105
Supplier:  MP Biomedicals
Acetate plate seal, MP Biomedicals
Catalog # ICNA097640105
Supplier:  MP Biomedicals
Specifications
  • Description:
    Acetate plate seal, 133×83 mm sheets
  • Pk:
    100
Specifications
About this item
Clear adhesive seals for multiwell plates and tissue culture plates.
  • Easy to apply
  • Suitable for both short and long term storage
  • Provides tight seal to minimise evaporation and condensation