Laboratory fume hoods, clear view, SP Bel-Art
BELAF500201010
- l×p×h ext.:657×610×663 mm
- Désignation:Laboratory hood, clear view, Scienceware®, model 1×1
- Cdt:1
- l×p×h int.:610×508×610 mm
Scienceware® Clear View fume hoods are suitable for potentially hazardous fumes and dust. It is constructed of polycarbonate with sleek modern style.
- Perfect for laboratory, classroom or industrial applications
- Visibility into the fume hood is possible from all sides
- Three position sliding sash can be completely lowered or set at a choice of two intermediary heights
- Opening (152 mm, O.D) at the top allows easy connection to exhaust systems