0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
ContactUsLinkComponent
Syringe Needles
Replacement needles for luer lock syringes, gas-tight (Hamilton)
Replacement needles for luer lock syringes, gas-tight (Hamilton)
Catalog # REST20133
Supplier:
Restek
Expand image
Replacement needles for luer lock syringes, gas-tight (Hamilton)
Catalog # REST20133
Supplier:
Restek
Specifications
Gauge:
26s
Type of tip:
2
Description:
Replacement needles, metal hub
Length:
51 mm
Seq:
05
Pk:
6
Specifications
About this item
Product Details
Product Details