Capillary columns, MXT®-1HT
Supplier: Restek
Jump to:
About this item
MXT®-1HT Siltek®-treated stainless steel nonpolar phase columns.
- Lowest bleed for longest column lifetime
- 100% dimethyl polysiloxane phase allows easy comparisons to historical data
- Individually tested
- 177,8 mm coil diameter
- Temperature ranges from 60 to 450 °C
: Meets all ASTM D2887, D6352, D7169, D7213, and D7500 specifications.
9 Options Available Below
Product Details & Documents
MXT®-1HT Siltek®-treated stainless steel nonpolar phase columns.
- Lowest bleed for longest column lifetime
- 100% dimethyl polysiloxane phase allows easy comparisons to historical data
- Individually tested
- 177,8 mm coil diameter
- Temperature ranges from 60 to 450 °C
: Meets all ASTM D2887, D6352, D7169, D7213, and D7500 specifications.
Product Family Options
Product Information
- Column FormatPhaseFilm thicknessØ int.LengthAvailabilityPrice