ContactUsLinkComponent
Glass microfibre filters with binder, grade GF 92, Whatman™
Glass microfibre filters with binder, grade GF 92, Whatman™
Certificates
undefined
Glass microfibre filters with binder, grade GF 92, Whatman™
Specifications
  • Catalog No:
  • 514-3816
  • 514-3817
  • 514-3819
  • 514-3818
  • 514-3821
  • 514-3814
  • Grade:
  • GF 92
  • GF 92
  • GF 92
  • GF 92
  • GF 92
  • GF 92
  • Weight:
  • 70 g/m²
  • 70 g/m²
  • 70 g/m²
  • 70 g/m²
  • 70 g/m²
  • 70 g/m²
  • Thickness:
  • 350 µm
  • 350 µm
  • 350 µm
  • 350 µm
  • 350 µm
  • 350 µm
  • Airflow rate:
  • 27 s/100 ml/1,56 cm²
  • 27 s/100 ml/1,56 cm²
  • 27 s/100 ml/1,56 cm²
  • 27 s/100 ml/1,56 cm²
  • 27 s/100 ml/1,56 cm²
  • 27 s/100 ml/1,56 cm²
  • Max. temperature:
  • 500 °C
  • 500 °C
  • 500 °C
  • 500 °C
  • 500 °C
  • 500 °C