Order Entry
Belgium
ContactUsLinkComponent
Application specific filter papers, grade 512 ½, Whatman™
Application specific filter papers, grade 512 ½, Whatman™
Certificates
undefined
Application specific filter papers, grade 512 ½, Whatman™

Specifications

  • Catalog No:
  • 512-0590
  • 512-0583
  • 515-5012
  • Grade:
  • 512 ½
  • 512 ½
  • 512 ½
  • Weight:
  • 84 g/m²
  • 84 g/m²
  • 84 g/m²
  • Thickness:
  • 160 µm
  • 160 µm
  • 160 µm
  • Filtration speed:
  • 375 s (Herzberg)
  • 375 s (Herzberg)
  • 375 s (Herzberg)

Specifications

Frequently Bought Together